missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Activin RIA/ALK-2 Fc Chimera Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 6506-RA-100
267.81 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
Measured by its binding ability in a functional ELISA. When Recombinant Mouse Activin RIA Fc Chimera is immobilized at 2 μg/mL (100 μL/well), the concentration of Recombinant Human BMP-6 (Catalog # 507-BP ) that produces 50% of the optimal binding response is found to be 6-60 ng/mL.Specifications
Lyophilized from a 0.2μm filtered solution in PBS. | |
Activin RIA/ALK-2 Fc Chimera | |
Mouse myeloma cell line,NS0-derived mouse Activin RIA/ALK-2 protein, (N-terminus) Mouse Activin RIA (Met1-Glu123) Accession &Num; P37172 IEGRMDP Mouse IgG2A (Glu98-Lys330) (C-terminus) | |
Disulfide-linked homodimer | |
Recombinant |
38.4 kDa (monomer) | |
100 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |