missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse aFGF / FGF1 Protein
Recombinant Protein
Brand: enQuireBio™ QP5455-500ug
This item is not returnable.
View return policy
Specifications
NP_034327.1 | |
aFGF / FGF1 Protein | |
Research Use Only | |
FGF1 | |
Recombinant Protein | |
E. coli | |
MFNLPLGNYK KPKLLYCSNG GHFLRILPDG TVDGTRDRSD QHIQLQLSAE SAGEVYIKGT ETGQYLAMDT EGLLYGSQTP NEECLFLERL EENHYNTYTS KKHAEKNWFV GLKKNGSCKR GPRTHYGQKA ILFLPLPVSS D | |
Greater than 95% as determined by SDS-PAGE. |
14164 | |
500 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Mouse aFGF / FGF1 induces 3T3 cell proliferation (typical ED50 is around 0.1 ng/ml). | |
Mouse | |
Untagged | |
10 mM sodium phosphate, 75 mM sodium chloride, pH 7.5 |