missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse Aminopeptidase PILS/ARTS1 Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Brand:  R&D Systems™ 2500-ZN-010

315.33 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18146120

  • £412.00 / 10µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Measured by its ability to cleave the fluorogenic peptide substrate, Leu-AMC. The specific activity is >100 pmol/min/μg, as measured under the described conditions.
Specifications

Specifications

Q6GTP5
Supplied as a 0.2 μm filtered solution in Tris, NaCl and Glycerol.
M.W. (Observed): 111 kDa, reducing conditions; M.W. (theoretical): 105 kDa
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Erap1
Recombinant
Aminopeptidase PILS/ARTS1
Enzyme Activity
51752
Aminopeptidase PILS/ARTS1
Mouse myeloma cell line, NS0-derived mouse Aminopeptidase PILS/ARTS1 protein Ser27-Leu930, with a C-terminal 10-His tag
<1.0 EU / 1 μg of the protein by the LAL method.
Unconjugated
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers