missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Beta-defensin 4 Protein
A DNA sequence encoding the Mus musculus (Mouse) Beta-defensin 4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6967-200ug
This item is not returnable.
View return policy
Specifications
NP_062702.1 | |
Beta-defensin 4 Protein | |
Research Use Only | |
DEFB4A | |
Recombinant Protein | |
E. coli | |
QIINNPITCM TNGAICWGPC PTAFRQIGNC GHFKVRCCKI R | |
Greater than 90% as determined by SDS-PAGE. |
56519 | |
200 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction