missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse BID (Caspase-8-cleaved) Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 883-M8-050

282.50 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18158020

  • £359.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Measured by its ability to induce cytochrome c release from isolated mouse liver mitochondria. The typical EC50 for cytochrome c releasing activity is <100 nM. Cytochrome c is quantified using the Rat/Mouse Cytochrome c Quantikine™ ELISA Kit (Catalog # MCTC0 ). The EC50 for the desired application should be determined. Uncleaved Recombinant Mouse BID is available (Catalog # 860-MB ).
Specifications

Specifications

P70444
Supplied as a 0.2 μm filtered solution in HEPES-KOH and KCl.
M.W. (Observed): 7 kDa and 15 kDa, under reducing and non-reducing conditions; M.W. (theoretical): 7 kDa (N-terminal fragment) & 15 kDa (C-terminal fragment)
50 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Unconjugated
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Bioactivity
637
BID Caspase-8-cleaved
E. coli-derived mouse BID protein Met1-Asp195
Bid
Recombinant
BID
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers