missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse BMP-7 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 5666-BP-010
This item is not returnable.
View return policy
Description
The Recombinant Mouse BMP-7 Protein is derived from CHO. The Recombinant Mouse BMP-7 Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein. | |
15.6 kDa (monomer) | |
10μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
12162 | |
BMP-7 | |
Chinese Hamster Ovary cell line,CHO-derived mouse BMP-7 protein Ser292-His430 | |
Disulfide-linked homodimer | |
Recombinant |