missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Camk2n1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Camk2n1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5751-ec-500ug
This item is not returnable.
View return policy
Specifications
NP_079727.1 | |
Camk2n1 Protein | |
Research Use Only | |
Camk2n1 | |
Recombinant Protein | |
E. coli | |
MSEVLPYGDE KLSPYGDGGD VGQIFSCRLQ DTNNFFGAGQ SKRPPKLGQI GRSKRVVIED DRIDDVLKTM TDKAPPGV | |
Greater than 90% as determined by SDS-PAGE. |
66259 | |
500 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |