Learn More
R&D Systems™ Recombinant Mouse CCL1/TCA-3 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand:  R&D Systems™ 845-TC-025/CF
 View more versions of this product
Product Code. 18157530
  • £396.00 / 25µg
Explore more special offers
Description
Description
Measured by its ability to chemoattract BaF3 mouse pro-B cells transfected with mouse CCR8. The ED50 for this effect is 0.5-2.0 ng/mL.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
7.8 kDa
25 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
20290
CCL1/I-309/TCA-3
Mouse myeloma cell line,NS0-derived mouse CCL1/I-309/TCA-3 protein Lys24-Cys92
Unconjugated
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain.