missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse CCL25/TECK Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

£361.00

Specifications

Gene ID (Entrez) 20300
Molecular Weight (g/mol) 14 kDa
Name CCL25/TECK
Quantity 25 μg
Source E. coli-derived mouse CCL25/TECK protein Gln24-Asn144,with an N-terminal Met
View More Specs

Products 2
Product Code Brand Formulation Price Quantity & Availability  
Product Code Brand Formulation Price Quantity & Availability  
18367737
View Documents
R&D Systems™
481-TK-025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
£361.00
25µg
Please to purchase this item. Need a web account? Register with us today!
18370088
View Documents
R&D Systems™
481-TK-025/CF
Lyophilized from a 0.2μm filtered solution in PBS.
£361.00
25µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Recombinant Mouse CCL25/TECK Protein is derived from E. coli. The Recombinant Mouse CCL25/TECK Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

20300
CCL25/TECK
E. coli-derived mouse CCL25/TECK protein Gln24-Asn144,with an N-terminal Met
Unconjugated
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
14 kDa
25 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
R&D Systems™ Recombinant Mouse CCL25/TECK Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.