missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse CCL4/MIP-1 beta Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£219.00 - £376.00
Specifications
Name | CCL4/MIP-1 beta |
---|---|
Source | E. coli-derived mouse CCL4/MIP-1 beta protein Ala24-Asn92 |
Conjugate | Unconjugated |
Recombinant | Recombinant |
Product Code | Brand | Quantity | Formulation | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Formulation | Price | Quantity & Availability | ||||
18116200
|
R&D Systems™
451-MB-050 |
50 μg | Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. |
£376.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18167490
|
R&D Systems™
451-MB-010 |
10 μg | Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. |
£219.00
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18328787
|
R&D Systems™
451-MB-010/CF |
10 μg | Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA with Trehalose. |
£219.00
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18147110
|
R&D Systems™
451-MB-050/CF |
50 μg | Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA with Trehalose. |
£376.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Measured by its ability to chemoattract BaF3 mouse pro-B cells transfected with human CCR5. The ED50 for this effect is 0.75-5 ng/mL.Specifications
CCL4/MIP-1 beta | |
E. coli-derived mouse CCL4/MIP-1 beta protein Ala24-Asn92 | |
Unconjugated | |
Recombinant |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title