missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse CD82 / KAI-1 Protein

A DNA sequence encoding the Mus musculus (Mouse) CD82 / KAI-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5799-ec-100ug

 View more versions of this product

Product Code. 15967849

  • £560.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_001129527.1
CD82 / KAI-1 Protein
Research Use Only
Cd82
Recombinant Protein
E. coli
DKLKKEMGNT VMDIIRNYTA NATSSREEAW DYVQAQVKCC GWVSHYNWTE NEELMGFTKT TYPCSCEKIK EEDNQLIVKK GFCEADNSTV SENNPEDWPV NTEGCMEKAQ AWLQENF
Greater than 90% as determined by SDS-PAGE.
12521
100 μg
Not determined.
Extracellular Domain
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers