missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CES1D Protein
A cDNA sequence encoding the CES1D was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10550-50ug
This item is not returnable.
View return policy
Specifications
CES1D Protein | |
Research Use Only | |
Specific activity is > 80,000 pmol/min/ug and is defined as the amount of enzyme that hydrolyze 1pmole of p-nitrophenyl acetate to pnitrophenol per minute at pH 7.5 at 37C. | |
Mouse | |
His | |
CES1D protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |
50 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
Baculovirus | |
YPSSPPVVNT VKGKVLGKYV NLEGFTQPVA VFLGVPFAKP PLGSLRFAPP QPAEPWSFVK NTTSYPPMCS QDAVGGQVLS ELFTNRKENI PLQFSEDCLY LNIYTPADLT KNSRLPVMVW IHGGGLVVGG ASTYDGLALS AHENVVVVTI QYRLGIWGFF STGDEHSRGN WGHLDQVAALRWVQDNIANF GGNPGSVTIF GESAGGFSVS VLVLSPLAKN LFHRAISESG VSLTAALITT DVKPIAGLVA TLSGCKTTTS AVMVHCLRQK TEDELLETSL KLNLFKLDLL GNPKESYPFL PTVIDGVVLP KAPEEILAEK SFSTVPYIVG INKQEFGWII PTLMGYPLAE GKLDQKTANSLLWKSYPTLK ISENMIPVVA EKYLGGTDDL TKKKDLFQDL MADVVFGVPS VIVSRSHRDA GASTYMYEFE YRPSFVSAMR PKAVIGDHGD EIFSVFGSPF LKDGASEEET NLSKMVMKFW ANFARNGNPN GGGLPHWPEY DQKEGYLKIG ASTQAAQRLK DKEVSFWAEL RAKESAQRPSHREHVELLEH HHHHH | |
Greater than 90.0% as determined by SDS-PAGE. |