Learn More
R&D Systems™ Recombinant Mouse Coagulation Factor III/Tissue Factor
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
Brand:  R&D Systems™ 3178-PA-010
Product Code. 18108680
  • £333.00 / 10µg
Explore more special offers
Description
Description
The Recombinant Mouse Coagulation Factor III/Tissue Factor is derived from NS0. The Recombinant Mouse Coagulation Factor III/Tissue Factor has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in Tris, NaCl and Brij-35.
26 kDa
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
14066
Coagulation Factor III/Tissue Factor
Mouse myeloma cell line,NS0-derived mouse Coagulation Factor III/Tissue Factor protein Ala29-Glu251,with a C-terminal 6-His tag
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
R&D Systems™ Recombinant Mouse Coagulation Factor III/Tissue Factor > 10ug; Carrier Free