missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Coagulation Factor VII Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
Brand: R&D Systems™ 3305-SE-010
This item is not returnable.
View return policy
Description
The Recombinant Mouse Coagulation Factor VII Protein is derived from CHO. The Recombinant Mouse Coagulation Factor VII Protein has been validated for the following applications: Enzyme Activity.Specifications
Lyophilized from a 0.2μm filtered solution in Tris and NaCl. | |
47 kDa | |
10 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
Reconstitute at 200 μg/mL in sterile 50 mM Tris, 10 mM CaCl 2 150 mM NaCl and 0.05% Brij-35 (pH 7.5). |
14068 | |
Coagulation Factor VII | |
Chinese Hamster Ovary cell line,CHO-derived mouse Coagulation Factor VII protein Ala42-Leu446,with a C-terminal 10-His tag | |
Mature form | |
Recombinant | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
R&D Systems™ Recombinant Mouse Coagulation Factor VII Protein > 10ug; Carrier Free
Spot an opportunity for improvement?Share a Content Correction