missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Complement C1q subcomponent subunit A Protein

A DNA sequence encoding the Mus musculus (Mouse) Complement C1q subcomponent subunit A, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5735-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15967379

  • £1600.00 / 500µg
Estimated Shipment: 29-08-2024
to see stock.

Description

Description

Specifications

Specifications

NP_031598.2
Complement C1q subcomponent subunit A Protein
Research Use Only
C1QA
Recombinant Protein
Yeast
EDVCRAPNGK DGAPGNPGRP GRPGLKGERG EPGAAGIRTG IRGFKGDPGE SGPPGKPGNV GLPGPSGPLG DSGPQGLKGV KGNPGNIRDQ PRPAFSAIRQ NPMTLGNVVI FDKVLTNQES PYQNHTGRFI CAVPGFYYFN FQVISKWDLC LFIKSSSGGQ PRDSLSFSNT NNKGLFQVLA GGTVLQLRRG DEVWIEKDPA KGRIYQGTEA DSIFSGFLIF PSA
Greater than 90% as determined by SDS-PAGE.
12259
500 μg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers