missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CXCL12 / SDF-1 Isoform beta Protein
Recombinant Protein
Brand: enQuireBio™ QP5216-2ug
This item is not returnable.
View return policy
Specifications
NP_038683.1 | |
2 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Mouse CXCL12 / SDF-1 Isoform beta induces primary human T cell chemotaxis (significanty different from control at Mouse CXCL12 / SDF-1 Isoform beta concentrations greater than 5-50 ng/ml). | |
Mouse | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
CXCL12 / SDF-1 Isoform beta Protein | |
Research Use Only | |
CXCL12 | |
Recombinant Protein | |
E. coli | |
KPVSLSYRCP CRFFESHIAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNKRL KM | |
Greater than 95% as determined by SDS-PAGE. |