missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse CXCL9/MIG Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

£236.00 - £358.00

Specifications

Gene ID (Entrez) 17329
Molecular Weight (g/mol) 12 kDa
Name CXCL9/MIG
Source E. coli-derived mouse CXCL9/MIG protein Thr22-Thr126
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
View More Specs

Products 4
Product Code Brand Quantity Formulation Price Quantity & Availability  
Product Code Brand Quantity Formulation Price Quantity & Availability  
18187940
View Documents Promotion Details
R&D Systems™
492-MM-010/CF
10 μg Lyophilized from a 0.2μm filtered solution in PBS.
£236.00
10µg
Please to purchase this item. Need a web account? Register with us today!
18108400
View Documents Promotion Details
R&D Systems™
492-MM-010
10 μg Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
£236.00
10µg
Please to purchase this item. Need a web account? Register with us today!
18126840
View Documents Promotion Details
R&D Systems™
492-MM-050
50 μg Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
£358.00
50µg
Please to purchase this item. Need a web account? Register with us today!
18128710
View Documents Promotion Details
R&D Systems™
492-MM-050/CF
50 μg Lyophilized from a 0.2μm filtered solution in PBS.
£358.00
50µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Recombinant Mouse CXCL9/MIG Protein is derived from E. coli. The Recombinant Mouse CXCL9/MIG Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

17329
CXCL9/MIG
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
12 kDa
E. coli-derived mouse CXCL9/MIG protein Thr22-Thr126
Unconjugated
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Videos
SDS
Documents

Documents

Product Certifications