missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Erythropoietin/EPO Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 959-ME-010/CF
316.19 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
The Recombinant Mouse Erythropoietin Protein is derived from NS0. The Recombinant Mouse Erythropoietin Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS. | |
18.6 kDa | |
10 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
>90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
13856 | |
Erythropoietin/EPO | |
Mouse myeloma cell line, NS0-derived mouse Erythropoietin/EPO protein Ala27-Arg192 | |
ECYT5, EP, EPO, epoetin, erythropoietin, MGC138142, MVCD2 | |
Recombinant |