missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Excitatory amino acid transporter 2 Protein

A DNA sequence encoding the Mus musculus (Mouse) Excitatory amino acid transporter 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6691-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16005150

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001070982.1
Excitatory amino acid transporter 2 Protein
Research Use Only
Slc1a2
Recombinant Protein
E. coli
HPGNPKLKKQ LGPGKKNDEV SSLDAFLDLI RNLFPENLVQ ACFQQIQTVT KKVLVAPPSE EANTTKAVIS MLNETMNEAP EETKIVIKKG LEFKDG
Greater than 90% as determined by SDS-PAGE.
20511
200 μg
Not determined.
Extracellular Domain
Mouse
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers