missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Excitatory amino acid transporter 2 Protein
A DNA sequence encoding the Mus musculus (Mouse) Excitatory amino acid transporter 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6691-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001070982.1 | |
Excitatory amino acid transporter 2 Protein | |
Research Use Only | |
Slc1a2 | |
Recombinant Protein | |
Yeast | |
HPGNPKLKKQ LGPGKKNDEV SSLDAFLDLI RNLFPENLVQ ACFQQIQTVT KKVLVAPPSE EANTTKAVIS MLNETMNEAP EETKIVIKKG LEFKDG | |
Greater than 90% as determined by SDS-PAGE. |
20511 | |
1 mg | |
Not determined. | |
Extracellular Domain | |
Mouse | |
GST | |
Tris-based buffer, 50% glycerol |