missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Excitatory amino acid transporter 2 Protein

A DNA sequence encoding the Mus musculus (Mouse) Excitatory amino acid transporter 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6691-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16075140

  • £560.00 / 100µg
Estimated Shipment: 29-08-2024
to see stock.

Description

Description

Specifications

Specifications

NP_001070982.1
Excitatory amino acid transporter 2 Protein
Research Use Only
Slc1a2
Recombinant Protein
E. coli
HPGNPKLKKQ LGPGKKNDEV SSLDAFLDLI RNLFPENLVQ ACFQQIQTVT KKVLVAPPSE EANTTKAVIS MLNETMNEAP EETKIVIKKG LEFKDG
Greater than 90% as determined by SDS-PAGE.
20511
100 μg
Not determined.
Extracellular Domain
Mouse
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers