missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse FABP1 Protein

A cDNA sequence encoding the FABP1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11829-25ug

 View more versions of this product

Product Code. 15958508

  • £155.00 / 25µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

FABP1 Protein
Research Use Only
Recombinant Protein
E. coli
MNFSGKYQLQ SQENFEPFMK AIGLPEDLIQ KGKDIKGVSE IVHEGKKIKL TITYGPKVVR NEFTLGEECE LETMTGEKVK AVVKLEGDNK MVTTFKGIKS VTELNGDTIT NTMTLGDIVY KRVSKRI
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
25 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Mouse
Untagged
Lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers