missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Glyoxylate reductase / hydroxypyruvate reductase Protein
A DNA sequence encoding the Mus musculus (Mouse) Glyoxylate reductase/hydroxypyruvate reductase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6119-ec-50ug
This item is not returnable.
View return policy
Specifications
NP_525028.1 | |
Glyoxylate reductase / hydroxypyruvate reductase Protein | |
Research Use Only | |
Glxr | |
Recombinant Protein | |
E. coli | |
MKPARLMKVF VTGPLPAEGR AALAQAADCE VEQWNSDDPI PRKDLEQGVV GAHGLLCRLS DRVDKKLLDA AGANLRVIST LSVGVDHLAL DEIKKRGIRV GYTPGVLTDA TAELAVSLLL TTCRRLPEAI EEVKNGGWSS WSPLWMCGYG LSQSTVGIVG LGRIGQAIAR RLKPFGVQRF LYTGRQPRPQ EAAEFQAEFV PIAQLAAESD FIVVSCSLTP DTMGLCSKDF FQKMKNTAIF INISRGDVVN QEDLYQALAS GQIAAAGLDV TTPEPLPPSH PLLTLKNCVI LPHIGSATYK TRNTMSLLAA NNLLAGLRGE AMPSELKL | |
Greater than 90% as determined by SDS-PAGE. |
76238 | |
50 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |