missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse GSTM1 Protein
A cDNA sequence encoding the GSTM1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10658-HIS-5ug
This item is not returnable.
View return policy
Specifications
GSTM1 Protein | |
Research Use Only | |
Specific activity is 3 units/mg, and is defined as the amount of enzyme that conjugate 1.0 u mole of 1-chloro-2,4-dinitrobenzene (CDNB) with reduced glutathione per minute at pH 6.5 at 25C. | |
Mouse | |
His | |
The GSTM1 solution contains PBS pH 7.4 and 5mM glutathione. |
5 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MPMILGYWNV RGLTHPIRML LEYTDSSYDE KRYTMGDAPD FDRSQWLNEK FKLGLDFPNL PYLIDGSHKI TQSNAILRYL ARKHHLDGET EEERIRADIV ENQVMDTRMQ LIMLCYNPDF EKQKPEFLKT IPEKMKLYSE FLGKRPWFAG DKVTYVDFLA YDILDQYRMF EPKCLDAFPN LRDFLARFEG LKKISAYMKS SRYIATPIFS KMAHWSNK | |
Greater than 95.0% as determined by SDS-PAGE. |