missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse HAI-1 Protein

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Inhibition Activity

Brand:  R&D Systems™ 1141-PI-010

261.77 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18158440

  • £333.00 / 10µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Recombinant Mouse HAI-1 Protein is derived from NS0. The Recombinant Mouse HAI-1 Protein has been validated for the following applications: Inhibition Activity.
Specifications

Specifications

Lyophilized from a 0.2 μm filtered solution in Tris, NaCl and CaCl 2 .
48 kDa
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
20732
HAI-1
Mouse myeloma cell line,NS0-derived mouse HAI-1 protein Glu30-Glu443 (Cys325Ser),with a C-terminal 10-His tag
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers