missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse Heparan Sulfate-6-O-Sulfotransferase-3

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Brand:  R&D Systems™ 5406-ST-020

303.23 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18136620

  • £385.00 / 20µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Measured by its ability to transfer sulfate from PAPS to heparan sulfate. The specific activity is >40 pmol/min/μg, as measured under the described conditions.
Specifications

Specifications

NP_056635
Supplied as a 0.2 μm filtered solution in Tris, NaCl and EDTA.
M.W. (Observed): 70-90 kDa, reducing conditions; M.W. (theoretical): 53 kDa
20 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Hs6st3
Recombinant
Heparan Sulfate 6-O-Sulfotransferase 3/HS6ST3
Enzyme Activity
266722
Heparan Sulfate 6-O-Sulfotransferase 3/HS6ST3
Chinese Hamster Ovary cell line, CHO-derived mouse Heparan Sulfate 6-O-Sulfotransferase 3/HS6ST3 protein Pro28-Trp470, with an N-terminal 6-His tag
<1.0 EU / 1 μg of the protein by the LAL method.
Unconjugated
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers