missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse HPD Protein
A cDNA sequence encoding the HPD was constructed and used to recombinantly synthesize the protein.
£99.15 - £1320.00
Specifications
Name | HPD Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Mouse |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15959978
|
enQuireBio™
QP12299-1ug |
1 μg |
£99.15
1µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15979978
|
enQuireBio™
QP12299-5ug |
5 μg |
£155.00
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15969978
|
enQuireBio™
QP12299-50ug |
50 μg |
£1320.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
HPD Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
HPD protein solution (0.5 mg/ml) contains 10% glycerol and 20mM Tris-HCl (pH 8.0). |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMTTYNNK GPKPERGRFL HFHSVTFWVG NAKQAASFYC NKMGFEPLAY RGLETGSREV VSHVIKQGKI VFVLCSALNP WNKEMGDHLV KHGDGVKDIA FEVEDCDHIV QKARERGAKI VREPWVEQDK FGKVKFAVLQ TYGDTTHTLV EKINYTGRFL PGFEAPTYKD TLLPKLPRCN LEIIDHIVGN QPDQEMQSAS EWYLKNLQFH RFWSVDDTQV HTEYSSLRSI VVTNYEESIK MPINEPAPGR KKSQIQEYVD YNGGAGVQHI ALKTEDIITA IRHLRERGTE FLAAPSSYYK LLRENLKSAK IQVKESMDVL EELHILVDYD EKGYLLQIFT KPMQDRPTLF LEVIQRHNHQ GFGAGNFNSL FKAFEEEQAL RGNLTDLEPN GVRSGM | |
Greater than 90% as determined by SDS-PAGE. |