missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse IL-27 p28/IL-30 Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 7430-ML-010/CF

 View more versions of this product

Product Code. 18166400

  • £305.00 / 10µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Recombinant Mouse IL-27 p28/IL-30 Protein is derived from NS0. The Recombinant Mouse IL-27 p28/IL-30 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in HEPES, NaCl and CHAPS.
Observed Molecular Wt: 25-35kDa, reducing conditions, Predicted Molecular Wt.: 24.9kDa
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
246779
IL-27 p28/IL-30
Mouse myeloma cell line,NS0-derived mouse IL-27 p28/IL-30 protein Phe29-Ser234,with a C-terminal 10-His tag
Unconjugated
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.