missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL17 / IL17A Protein
Recombinant Protein
Brand: enQuireBio™ QP5466-25ug
This item is not returnable.
View return policy
Specifications
NP_034682.1 | |
IL17 / IL17A Protein | |
Research Use Only | |
IL17A | |
Recombinant Protein | |
E. coli | |
MAAIIPQSSA CPNTEAKDFL QNVKVNLKVF NSLGAKVSSR RPSDYLNRST SPWTLHRNED PDRYPSVIWE AQCRHQRCVN AEGKLDHHMN SVLIQQEILV LKREPESCPF TFRVEKMLVG VGCTCVASIV RQAA | |
Greater than 95% as determined by SDS-PAGE. |
16171 | |
25 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 5 ng/mL in the Production of IL-6 by mouse 3T3 cells assay. The bioactivity acceptance criteria is set at 2 x 10^5 units/mg. | |
Mouse | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |