missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL1RL2 / IL-1Rrp2 Protein
A DNA sequence encoding the Mus musculus (Mouse) IL1RL2 / IL-1Rrp2, was expressed in the hosts and tags indicated.
£281.57 - £2480.00
Specifications
Accession Number | NP_573456.1 |
---|---|
Gene ID (Entrez) | 107527 |
Name | IL1RL2 / IL-1Rrp2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16062851
|
enQuireBio™
QP8137-ec-10ug |
10 μg | |||||||
16002861
|
enQuireBio™
QP8137-ec-50ug |
50 μg | |||||||
16052851
|
enQuireBio™
QP8137-ec-100ug |
100 μg |
£715.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16082851
|
enQuireBio™
QP8137-ec-200ug |
200 μg | |||||||
16092851
|
enQuireBio™
QP8137-ec-500ug |
500 μg | |||||||
16072851
|
enQuireBio™
QP8137-ec-1mg |
1 mg |
£2480.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_573456.1 | |
IL1RL2 / IL-1Rrp2 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
Tris-based buffer, 50% glycerol |
107527 | |
Research Use Only | |
Il1rl2 | |
Recombinant Protein | |
DTCEDIFMHN VIISEGQPFP FNCTYPPETN GAVNLTWYKT PSKSPVSNNR HLRVHQDQTW ILFLPLTLED SGIYQCVIRN AHNCYQIAVN LTVLKNHWCD SSMEGSPVNS PDVYQQILPI GKSGSLNCHL YFPESCALDS IKWYKGCEEI KAGKKYSPSG AKLLVNNVAV EDGGSYACSA RLTHLGRHFT IRNYIAVNTK EVEYGRRIPN ITYPKNNSIE VPLGSTLIVN CNITDTKENT NLRCWRVNNT LVDDYYKDSK RIQEGIETNV SLRDQIRYTV NITFLKVKME DYGRPFTCHA GVSAAYIILI YPVPDFR |