missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse IL7R / IL7RA Protein

A DNA sequence encoding the Mus musculus (Mouse) IL7R / IL7RA, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6226-ba-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16081730

  • £476.00 / 20µg
Estimated Shipment: 05-09-2024
to see stock.

Description

Description

Specifications

Specifications

NP_032398.3
IL7R / IL7RA Protein
Research Use Only
Il7r
Recombinant Protein
Baculovirus
ESGNAQDGDL EDADADDHSF WCHSQLEVDG SQHLLTCAFN DSDINTANLE FQICGALLRV KCLTLNKLQD IYFIKTSEFL LIGSSNICVK LGQKNLTCKN MAINTIVKAE APSDLKVVYR KEANDFLVTF NAPHLKKKYL KKVKHDVAYR PARGESNWTH VSLFHTRTTI PQRKLRPKAM YEIKVRSIPH NDYFKGFWSE WSPSSTFETP EPKNQGGWD
Greater than 90% as determined by SDS-PAGE.
16197
20 μg
Not determined.
Extracellular Domain
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers