missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse IMPAD1 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 7028-PD-050
276.45 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
Measured by its ability to dephosphorylate 3'-Phosphoadenosine 5'-phosphate. The specific activity is >800 pmol/min/ug, as measured under the described conditions.Specifications
Q80V26 | |
Supplied as a 0.2 μm filtered solution in Tris, NaCl and Glycerol. | |
M.W. (Observed): 35-40 kDa, reducing conditions; M.W. (theoretical): 34 kDa | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. | |
Impad1 | |
Recombinant | |
Inositol Monophosphatase 3/IMPAD1 |
Enzyme Activity | |
54928 | |
Inositol Monophosphatase 3/IMPAD1 | |
Chinese Hamster Ovary cell line, CHO-derived mouse Inositol Monophosphatase 3/IMPAD1 protein Glu51-His356 with an N-terminal 6-His tag | |
<1.0 EU / 1 μg of the protein by the LAL method. | |
Unconjugated | |
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |