missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Integrin alpha 10 beta 1 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 7827-AB-050
This item is not returnable.
View return policy
Description
The Recombinant Mouse Integrin alpha 10 beta 1 Protein is derived from CHO. The Recombinant Mouse Integrin alpha 10 beta 1 Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS. | |
Integrin alpha 10 beta 1 | |
Chinese Hamster Ovary cell line,CHO-derived mouse Integrin alpha 10 beta 1 protein, (N-terminus) Mouse Integrin alpha 10 Phe23-Thr1118 (Lys ins between Gln858 and Ala859) Accession &Num; NP_001289400.1 His-Pro GGGSGGGS Acidic Tail HHHHHH Mouse Integrin beta 1 (Gln21-Asp728) Accession &Num; P09055 His-Pro GGGSGGGS Basic Tail (C-terminus) | |
Noncovalently-linked heterodimer | |
Recombinant |
129.2 kDa (Integrin alpha10) & 86.5 kDa (Integrin beta1) | |
50μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction