missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Integrin alpha 7 beta 1 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 7958-A7-050
261.77 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
Measured by its binding ability in a functional ELISA. When mouse Laminin I (Catalog # 3400-010-02 ) is coated at 10 µg/mL, Recombinant Mouse Integrin alpha 7 beta 1 binds with an apparent KD <0.5 nM.Specifications
Lyophilized from a 0.2μm filtered solution in PBS. | |
Integrin alpha 7 beta 1 | |
Chinese Hamster Ovary cell line,CHO-derived mouse Integrin alpha 7 beta 1 protein, (N-terminus) Mouse Integrin alpha 7 (Phe34-Glu1033) Accession &Num; NP_032424 His-Pro GGGSGGGS Acidic Tail 6-His tag Mouse Integrin beta 1 (Gln21-Asp728) Accession &Num; P09055 His-Pro GGGSGGGS Basic Tail (C-terminus) | |
Noncovalently-linked heterodimer | |
Recombinant |
119 kDa (Integrin alpha7, full length), 22.4 kDa (Integrin alpha7, N-terminus starts at Glu915) & 86.5 kDa (Integrin beta1) | |
50μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |