missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Integrin alpha L beta 2 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 7825-AB-050
282.50 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
The Recombinant Mouse Integrin alpha L beta 2 Protein is derived from CHO. The Recombinant Mouse Integrin alpha L beta 2 Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS. | |
Integrin alpha L beta 2 | |
Chinese Hamster Ovary cell line,CHO-derived mouse Integrin alpha L beta 2 protein, (N-terminus) Mouse Integrin alpha L (Tyr24-Glu1084) Accession &Num; P24063 His-Pro GGGSGGGS Acidic Tail 6-His tag Mouse Integrin beta 2 (Gln24-Asn702) Accession &Num; P11835 His-Pro GGGSGGGS Basic Tail (C-terminus) | |
Noncovalently-linked heterodimer | |
Recombinant |
126 kDa (Integrin alphaL) & 83 kDa (Integrin beta2) | |
50μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |