missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse Integrin alpha V beta 3 Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 7889-AV-050

261.77 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18147890

  • £343.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Recombinant Mouse Integrin alpha V beta 3 Protein is derived from CHO. The Recombinant Mouse Integrin alpha V beta 3 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
Integrin alpha V beta 3
Chinese Hamster Ovary cell line,CHO-derived mouse Integrin alpha V beta 3 protein, (N-terminus) Mouse Integrin alpha V (Phe31-Val988) Accession &Num; P43406 His-Pro GGGSGGGS Acidic Tail 6-His tag Mouse Integrin beta 3 (Glu26-Asp717) Accession &Num; O54890 His-Pro GGGSGGGS Basic Tail (C-terminus)
Noncovalently-linked heterodimer
Recombinant
115 kDa (Integrin alphaV) & 84.5 kDa (Integrin beta3)
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
90%, by SDS-PAGE with silver staining.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers