missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse LBP Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£343.00
Specifications
Gene ID (Entrez) | 16803 |
---|---|
Molecular Weight (g/mol) | 51.5 kDa |
Name | LBP |
Quantity | 25 μg |
Source | Mouse myeloma cell line,NS0-derived mouse LBP protein Gly25-Val481 (Ser102Arg,Tyr284His),with a C-terminal 6-His tag |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18128690
|
R&D Systems™
6635-LP-025/CF |
Lyophilized from a 0.2μm filtered solution in MES, NaCl, PEG and CHAPS. |
£343.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18166660
|
R&D Systems™
6635-LP-025 |
Lyophilized from a 0.2μm filtered solution in MES, NaCl, PEG and CHAPS with BSA as a carrier protein. |
£343.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Mouse LBP Protein is derived from NS0. The Recombinant Mouse LBP Protein has been validated for the following applications: Bioactivity.Specifications
16803 | |
LBP | |
Mouse myeloma cell line,NS0-derived mouse LBP protein Gly25-Val481 (Ser102Arg,Tyr284His),with a C-terminal 6-His tag | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
51.5 kDa | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title