missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse LIX Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£291.57
Specifications
For Use With (Application) | Bioactivity |
---|---|
Gene ID (Entrez) | 20311 |
Molecular Weight (g/mol) | 8 kDa |
Name | LIX |
Quantity | 25μg |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18168670
|
R&D Systems™
433-MC-025/CF |
Lyophilized from a 0.2μm filtered solution in PBS. |
£291.57
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18116420
|
R&D Systems™
433-MC-025 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
£291.57
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Mouse LIX Protein is derived from E. coli. The Recombinant Mouse LIX Protein has been validated for the following applications: Bioactivity.Specifications
Bioactivity | |
8 kDa | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
20311 | |
LIX | |
E. coli-derived mouse LIX protein Val45-Ala118 | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |