missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Lymphotoxin beta R/TNFRSF3 Fc Chimera
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 1008-LR-050
276.45 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
The Recombinant Mouse Lymphotoxin beta R/TNFRSF3 Fc Chimera is derived from NS0. The Recombinant Mouse Lymphotoxin beta R/TNFRSF3 Fc Chimera has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS. | |
48.7 kDa (monomer) | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
17000 | |
Lymphotoxin beta R/TNFRSF3 Fc Chimera | |
Mouse myeloma cell line,NS0-derived mouse Lymphotoxin beta R/TNFRSF3 protein, (N-terminus) Mouse Lymphotoxin beta R (Ser28-Pro218) Accession &Num; P50284 IEGRMDP Mouse IgG2A (Glu98-Lys330) (C-terminus) | |
Disulfide-linked homodimer | |
Recombinant |