missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse MAdCAM-1 Fc Chimera Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 993-MC-050
This item is not returnable.
View return policy
Description
The Recombinant Mouse MAdCAM-1 Fc Chimera Protein is derived from NS0. The Recombinant Mouse MAdCAM-1 Fc Chimera Protein has been validated for the following applications: Bioactivity.![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Lyophilized from a 0.2μm filtered solution in PBS. | |
63 kDa (monomer) | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
17123 | |
MAdCAM-1 Fc Chimera | |
Mouse myeloma cell line,NS0-derived mouse MAdCAM-1 protein, (N-terminus) Mouse MAdCAM-1 (Gln22-Thr365) Accession &Num; NP_038619 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) | |
Disulfide-linked homodimer | |
Recombinant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
R&D Systems™ Recombinant Mouse MAdCAM-1 Fc Chimera Protein > 50ug; Carrier Free
Spot an opportunity for improvement?Share a Content Correction