missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse MCP-2 / CCL8 Protein
Recombinant Protein
Brand: enQuireBio™ QP5471-500ug
This item is not returnable.
View return policy
Specifications
NP_067418.1 | |
MCP-2 / CCL8 Protein | |
Research Use Only | |
Ccl8 | |
Recombinant Protein | |
E. coli | |
GPDKAPVTCC FHVLKLKIPL RVLKSYERIN NIQCPMEAVV FQTKQGMSLC VDPTQKWVSE YMEILDQKSQ ILQP | |
Greater than 95% as determined by SDS-PAGE. |
20307 | |
500 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
THP-1 chemotaxis significantly induced by less than 100 ng/ml of Mouse MCP-2 / CCL8. | |
Mouse | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |