missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse MOG Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 8536-MO-050
261.77 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
The Recombinant Mouse MOG Protein is derived from NS0. The Recombinant Mouse MOG Protein has been validated for the following applications: Bioactivity.Specifications
Q61885 | |
4340 | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
MGC26137, MOG Ig-AluB, MOGIG2, myelin oligodendrocyte glycoprotein, myelin-oligodendrocyte glycoprotein | |
Mog | |
Unconjugated | |
Recombinant | |
>90%, by SDS-PAGE with Silver Staining |
Bioactivity | |
Observed M.W.: 19-29kDa, reducing conditions, Theoretical M.W.: 16kDa | |
Mouse myeloma cell line,NS0-derived mouse MOG protein Gly29-Gly153,with a C-terminal 10-His tag | |
<0.10 EU per 1μg of the protein by the LAL method. | |
MOG | |
Measured by its ability to enhance neurite outgrowth of E16-E18 rat embryonic cortical neurons. Recombinant Mouse MOG, immobilized at 1μg/mL on a 96-well plate, is able to significantly enhance neurite outgrowth. | |
Mouse | |
Mouse myeloma cell line, NS0-derived Gly29-Gly153, with a C-terminal 10-His tag |