missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse MUC5AC Protein
A DNA sequence encoding the Mus musculus (Mouse) MUC5AC, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8891-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_034974.1 | |
MUC5AC Protein | |
Research Use Only | |
Muc5ac | |
Recombinant Protein | |
E. coli | |
CPKNSSLIVT YEEGACCPTQ NCSSQKGCEV NGTLYQPGDV VSSSLCERCL CEVSSNPLSD VFMVSCETEL CNTQCPKGSE YQAMPGQCCG KCIPKTCPFK NNSGSTYFYQ PGELWAEPGN PCVTHKCEKF QDVLMVVTMK TECPKINCPQ GQAQLREDGC CYDCPLPNQQ KCTVHQRQQI IRQQNCSSEG PVSISYCQGN CGDSISMYSL EANKVEHTCE CCQELQTSQR NVTLRCDDGS SQTFSYTQVE KCGCLGQQCH ALGDTSHAES | |
Greater than 90% as determined by SDS-PAGE. |
17833 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |