missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse MuSK Fc Chimera Protein
Brand: R&D Systems™ 10599-MK-050
388.76 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
Measured by its binding ability in a functional ELISA. In the presence of Recombinant Mouse Agrin, immobilized Recombinant Mouse MuSK Fc Chimera (2 μg/mL at 100 μL/well) binds Recombinant Mouse LRP-4 (Catalog # 10229-LR ) with an ED50 of 150-900 ng/mLSpecifications
Q61006.1 | |
Bioactivity | |
50 μg | |
EC 2.7.10, EC 2.7.10.1, MGC126323, MGC126324, muscle, skeletal, receptor tyrosine kinase, MuSK, skeletal receptor tyrosine-protein kinase | |
Lyophilized from a 0.2 μm filtered solution in PBS. |
Unconjugated | |
4593 | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 2 weeks, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. | |
Mouse |