missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse NNMT Protein

A cDNA sequence encoding the NNMT was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12862-100ug

 View more versions of this product

Product Code. 15941779

  • £1320.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

18113
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMESGFT SKDTYLSHFN PRDYLEKYYS FGSRHCAENE ILRHLLKNLF KIFCLGAVKG ELLIDIGSGP TIYQLLSACE SFTEIIVSDY TDQNLWELQK WLKKEPGAFD WSPVVTYVCD LEGNRMKGPE KEEKLRRAIK QVLKCDVTQS QPLGGVSLPP ADCLLSTLCL DAACPDLPAY RTALRNLGSL LKPGGFLVMV DALKSSYYMI GEQKFSSLPL GWETVRDAVE EAGYTIEQFE VISQNYSSTT SNNEGLFSLV GRKPGRSE
Greater than 90.0% as determined by SDS-PAGE.
NNMT Protein
Research Use Only
Nnmt
Mouse
His
NNMT protein solution (1 mg/ml) containing 20mM Tris-HCl (pH 8.0), 10% glycerol and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers