missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse NUCB2 Protein
A cDNA sequence encoding the NUCB2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12896-5ug
This item is not returnable.
View return policy
Specifications
NUCB2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MVPIDVDKTK VHNTEPVENA RIEPPDTGLY YDEYLKQVIE VLETDPHFRE KLQKADIEEI RSGRLSQELD LVSHKVRTRL DELKRQEVGR LRMLIKAKLD ALQDTGMNHH LLLKQFEHLN HQNPNTFESR DLDMLIKAAT ADLEQYDRTR HEEFKKYEMMKEHERREYLK TLSEEKRKEE ESKFEEMKRK HEDHPKVNHP GSKDQLKEVW EETDGLDPND FDPKTFFKLH DVNNDGFLDE QELEALFTRE LEKVYNPQNA EDDMIEMEEE RLRMREHVMS EIDNNKDRLV TLEEFLRATE KKEFLEPDSW ETLDQQQLFT EDELKEYESI IAIQENELKKRAEELQKQKE DLQRQHDHLE AQKQEYHQAV QHLEQKKLQQ GIAPSGPAGE LKFEPHT | |
Greater than 95.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
The protein solution (1 mg/ml) contains 20mM Tris-HCl pH 8.0, 0.1M NaCl and 1mM EDTA. |