missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse PDGF-CC Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£343.00
Specifications
Gene ID (Entrez) | 54635 |
---|---|
Molecular Weight (g/mol) | 13.4 kDa (monomer) |
Name | PDGF-CC |
Quantity | 25 μg |
Source | E. coli-derived mouse PDGF-CC protein Val235-Gly345,with an N-terminal Met and a 6-His tag |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18300078
|
R&D Systems™
1447-PC-025 |
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. |
£343.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18196450
|
R&D Systems™
1447-PC-025/CF |
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. |
£343.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Mouse PDGF-CC Protein is derived from E. coli. The Recombinant Mouse PDGF-CC Protein has been validated for the following applications: Bioactivity.Specifications
54635 | |
PDGF-CC | |
E. coli-derived mouse PDGF-CC protein Val235-Gly345,with an N-terminal Met and a 6-His tag | |
Disulfide-linked homodimer | |
Recombinant |
13.4 kDa (monomer) | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |