Learn More
R&D Systems™ Recombinant Mouse PDGF R alpha Fc Chimera Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand:  R&D Systems™ 1062-PR-050
282.50 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18107550
  • £359.00 / 50µg
Explore more special offers
Description
Description
The Recombinant Mouse PDGF R alpha Fc Chimera Protein is derived from NS0. The Recombinant Mouse PDGF R alpha Fc Chimera Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS.
82.5 kDa (monomer)
50 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
18595
PDGFR alpha
Mouse myeloma cell line,NS0-derived mouse PDGF R alpha protein, (N-terminus) Mouse PDGF R alpha Leu25-Glu524 (Asp65Glu,Gly439Ala,Thr440Ala) Accession &Num; P26618.3 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Disulfide-linked homodimer
Recombinant
Product Suggestions