Learn More
R&D Systems™ Recombinant Mouse Pentraxin 3/TSG-14 Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity
Brand:  R&D Systems™ 2166-TS-025/CF
 View more versions of this product
Product Code. 18197910
  • £345.56 / 25µg
Explore more special offers
Description
Description
The Recombinant Mouse Pentraxin 3/TSG-14 Protein is derived from NS0. The Recombinant Mouse Pentraxin 3/TSG-14 Protein has been validated for the following applications: Binding Activity.
Specifications
Specifications
Lyophilized from a 0.2 μm filtered solution in MOPS, NaCl and CaCl 2 .
41 kDa (monomer)
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
19288
Pentraxin 3/TSG-14
Mouse myeloma cell line,NS0-derived mouse Pentraxin 3/TSG-14 protein Glu18-Ser381,with a C-terminal 6-His tag
Multimer consisting of as many as ten non-covalently and covalently linked subunits
Recombinant
Product Suggestions