missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse RSPO1 Protein
A cDNA sequence encoding the RSPO1 was constructed and used to recombinantly synthesize the protein.
£99.15 - £1320.00
Specifications
Gene ID (Entrez) | 192199 |
---|---|
Name | RSPO1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | Rspo1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15963339
|
enQuireBio™
QP13365-2ug |
2 μg |
£99.15
2µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15953339
|
enQuireBio™
QP13365-10ug |
10 μg |
£155.00
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15943339
|
enQuireBio™
QP13365-100ug |
100 μg |
£1320.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
192199 | |
Research Use Only | |
Rspo1 | |
Mouse | |
His | |
RSPO1 protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |
RSPO1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
ADPSRGIKGK RQRRISAEGS QACAKGCELC SEVNGCLKCS PKLFILLERN DIRQVGVCLP SCPPGYFDAR NPDMNKCIKC KIEHCEACFS HNFCTKCQEG LYLHKGRCYP ACPEGSTAAN STMECGSPAQ CEMSEWSPWG PCSKKRKLCG FRKGSEERTR RVLHAPGGDH TTCSDTKETR KCTVRRTPCP EGQKRRKGGQ GRRENANRHP ARKNSKEPGS NSRRHKGQQQ PQPGTTGPLT SVGPTWAQHH HHHH | |
Greater than 90.0% as determined by SDS-PAGE. |